accessibility menu, dialog, popup

UserName

SNNOCl

CAS RN 184970-24-9

IUPAC Name: 3-(chloromethyl)-5-(thiophen-3-yl)-1,2,4-oxadiazole
Synonyms: 3-(chloromethyl)-5-(thiophen-3-yl)-1,2,4-oxadiazole
Molecular Weight (g/mol): 200.64
Molecular Formula: C7H5ClN2OS
InChi Key: KHJYOJVUQYWASE-UHFFFAOYSA-N
SMILES: ClCC1=NOC(=N1)C1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results