accessibility menu, dialog, popup

UserName

CAS RN 1852-04-6

HOOOHO

CAS RN 1852-04-6

IUPAC Name: undecanedioic acid
Synonyms: undecanedioic acid
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: LWBHHRRTOZQPDM-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results