CAS RN 18520-63-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Monobenzyl Terephthalate, TRC
CAS: 18520-63-3 Molecular Formula: C15H12O4 Molecular Weight (g/mol): 256.25 Synonym: 1,4-Benzenedicarboxylic Acid Mono(phenylmethyl) Ester,Terephthalic Acid Benzyl Ester,Terephthalic Acid Monobenzyl Ester,Monobenzyl 1,4-Benzenedicarboxylate,Monobenzyl Terephthalate,p-(Benzyloxycarbonyl)benzoic Acid SMILES: COc1cc2c(cc1)c3c(c(C)ncc3)n2