accessibility menu, dialog, popup

UserName

PPdBrBrPdPH3CH3CCH3H3CH3CH3CH3CCH3CH3H3CCH3CH3CH3CH3CH3H3CCH3CH3

CAS RN 185812-86-6

IUPAC Name: tri-tert-butylphosphane palladium bromide
Synonyms: tri-tert-butylphosphane palladium bromide
Molecular Weight (g/mol): 388.65
Molecular Formula: C12H27BrPPd
InChi Key: WIADNXKBAGKNOT-UHFFFAOYSA-M
SMILES: [Br-].[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)