accessibility menu, dialog, popup

UserName

ClH3NNOO

CAS RN 18600-42-5

IUPAC Name: (4-nitrophenyl)methanaminium chloride
Synonyms: (4-nitrophenyl)methanaminium chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: SMIXZZMSWYOQPW-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results