accessibility menu, dialog, popup

UserName

CAS RN 1866-16-6

IUPAC Name: [2-(butanoylsulfanyl)ethyl]trimethylazanium iodide
Synonyms: butyrylthiocholine iodide
Molecular Weight (g/mol): 317.23
Molecular Formula: C9H20INOS
InChi Key: WEQAAFZDJROSBF-UHFFFAOYSA-M
SMILES: [I-].CCCC(=O)SCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)