CAS RN 18694-40-1
CAS RN 18694-40-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mepirizole, TRC
CAS: 18694-40-1 Molecular Formula: C11H14N4O2 Molecular Weight (g/mol): 234.25 Synonym: 4-Methoxy-2-(5-methoxy-3-methylpyrazol-1-yl)-6-methyl-pyrimidine,4-Methoxy-2-(5-methoxy-3-methyl-1H-pyrazol-1-yl)-6-methylpyrimidine,1-(4-Methoxy-6-methyl-2-pyrimidinyl)-3-methyl-5-methoxypyrazole,1-(4-Methyl-6-methoxy-2-pyrimidinyl)-3-methyl-5-methoxypyrazole,2-(3-Methoxy-5-methylpyrazol-2-yl)-4-methoxy-6-methylpyrimidine,2-(3-Methyl-5-methoxy-1-pyrazolyl)-4-methoxy-6-methylpyrimidine,4-Methoxy-2-(5-methoxy-3-methylpyrazol-1-yl)-6-methylpyrimidine,DA 398,Epirizole,Mebron SMILES: COc1cc(C)nc(n1)n2nc(C)cc2OC