accessibility menu, dialog, popup

UserName

SbH3CH3COCH3CH3OCH3CH3O

CAS RN 18770-47-3

IUPAC Name: antimony(3+) tris(propan-2-olate)
Synonyms: antimony(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 299.02
Molecular Formula: C9H21O3Sb
InChi Key: HYPTXUAFIRUIRD-UHFFFAOYSA-N
SMILES: [Sb+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)