accessibility menu, dialog, popup

UserName

MnS

CAS RN 18820-29-6

IUPAC Name: manganese sulfane
Synonyms: manganese(II) sulfide
Molecular Weight (g/mol): 89.01
Molecular Formula: H2MnS
InChi Key: TWJXYBSUGSKHPM-UHFFFAOYSA-N
SMILES: S.[Mn]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results