accessibility menu, dialog, popup

UserName

CAS RN 188844-34-0

H3CNNOONOOHCH3CH3H3CCH3

CAS RN 188844-34-0

IUPAC Name: 4-{2,14,14,17,17-pentamethyl-6-nitro-2,9-diazatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(19),3(8),4,6,9,11,13(18)-heptaen-10-yl}benzoic acid
Synonyms: 4-{2,14,14,17,17-pentamethyl-6-nitro-2,9-diazatetracyclo[9.8.0.0³,⁸.0¹³,¹⁸]nonadeca-1(19),3(8),4,6,9,11,13(18)-heptaen-10-yl}benzoic acid
Molecular Weight (g/mol): 483.57
Molecular Formula: C29H29N3O4
InChi Key: SXKPGYKPQPYJER-UHFFFAOYSA-N
SMILES: CN1C2=CC3=C(C=C2C(=NC2=C1C=CC(=C2)[N+]([O-])=O)C1=CC=C(C=C1)C(O)=O)C(C)(C)CCC3(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results