CAS RN 18972-56-0
CAS RN 18972-56-0
IUPAC Name:
silicon(4+) magnesium(2+) hexahydrate hexafluoride
Synonyms:
silicon(4+) magnesium(2+) hexahydrate hexafluoride
Molecular Weight (g/mol):
274.47
Molecular Formula:
F6H12MgO6Si
InChi Key:
QNZBQSBPLQILSY-UHFFFAOYSA-H
SMILES:
O.O.O.O.O.O.[F-].[F-].[F-].[F-].[F-].[F-].[Mg++].[Si+4]
1
–
2
of
2
results
1
Magnesium hexafluorosilicate hexahydrate, 98%
CAS: 18972-56-0 Molecular Formula: MgSiF6·6H2O MDL Number: MFCD00150250