accessibility menu, dialog, popup

UserName

ONONHNNOONOO

CAS RN 1898-66-4

IUPAC Name: 1,1-diphenyl-2-(2,4,6-trinitrophenyl)hydrazine
Synonyms: DPPH
Molecular Weight (g/mol): 395.33
Molecular Formula: C18H13N5O6
InChi Key: WCBPJVKVIMMEQC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=C(NN(C2=CC=CC=C2)C2=CC=CC=C2)C(=C1)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)