accessibility menu, dialog, popup

UserName

H3COOH3CCH3CH3(E/Z)

CAS RN 190059-33-7

IUPAC Name: 4-{1,3,3-trimethyl-7-oxabicyclo[4.1.0]heptan-2-yl}but-3-en-2-one
Synonyms: 4-{1,3,3-trimethyl-7-oxabicyclo[4.1.0]heptan-2-yl}but-3-en-2-one
Molecular Weight (g/mol): 208.3
Molecular Formula: C13H20O2
InChi Key: ODMUHAHUBCUABS-UHFFFAOYNA-N
SMILES: CC(=O)C=CC1C2(C)OC2CCC1(C)C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results