accessibility menu, dialog, popup

UserName

CH3ClNNCl

CAS RN 19064-64-3

IUPAC Name: 3,6-dichloro-4-methylpyridazine
Synonyms: 3,6-dichloro-4-methylpyridazine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: ROYHWGZNGMXQEU-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=NN=C1Cl
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)