accessibility menu, dialog, popup

UserName

CAS RN 190771-22-3

CH3SiH3CCH3N

CAS RN 190771-22-3

IUPAC Name: 3-[2-(trimethylsilyl)ethynyl]benzonitrile
Synonyms: 3-[2-(trimethylsilyl)ethynyl]benzonitrile
Molecular Weight (g/mol): 199.33
Molecular Formula: C12H13NSi
InChi Key: QEZDMZVLNWSJSA-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC(=CC=C1)C#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)