accessibility menu, dialog, popup

UserName

OO

CAS RN 191-28-6

IUPAC Name: 12,22-dioxahexacyclo[11.7.1.1⁴,²⁰.0²,¹¹.0³,⁸.0¹⁷,²¹]docosa-1(21),2,4,6,8,10,13,15,17,19-decaene
Synonyms: dinaphthylene dioxide
Molecular Weight (g/mol): 282.3
Molecular Formula: C20H10O2
InChi Key: AMDQVKPUZIXQFC-UHFFFAOYSA-N
SMILES: O1C2=CC=CC3=CC=C4OC5=CC=CC6=CC=C1C(=C56)C4=C23

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results