accessibility menu, dialog, popup

UserName

CH3NONO

CAS RN 19164-41-1

IUPAC Name: 5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate
Synonyms: 5-methyl-2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Weight (g/mol): 150.14
Molecular Formula: C7H6N2O2
InChi Key: HCWVKDOCXDWFEH-UHFFFAOYSA-N
SMILES: CC1=CC2=NO[N+]([O-])=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results