accessibility menu, dialog, popup

UserName

CH3H3CCH3OONNHNH3

CAS RN 192130-34-0

IUPAC Name: 1-(2-azaniumylethyl)-4-[(tert-butoxy)carbonyl]piperazin-1-ium
Synonyms: 1-(2-ammonioethyl)-4-(tert-butoxycarbonyl)piperazin-1-ium
Molecular Weight (g/mol): 231.34
Molecular Formula: C11H25N3O2
InChi Key: QSYTWBKZNNEKPN-UHFFFAOYSA-P
SMILES: CC(C)(C)OC(=O)N1CC[NH+](CC[NH3+])CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results