accessibility menu, dialog, popup

UserName

CH3CH3NCS

CAS RN 19241-15-7

IUPAC Name: 2-isothiocyanato-1,4-dimethylbenzene
Synonyms: 2-isothiocyanato-1,4-dimethylbenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: RCBXZRJVMSWGAO-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)N=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)