accessibility menu, dialog, popup

UserName

BClClFFFK

CAS RN 192863-38-0

IUPAC Name: potassium (2,4-dichlorophenyl)trifluoroboranuide
Synonyms: potassium (2,4-dichlorophenyl)trifluoroboranuide
Molecular Weight (g/mol): 252.89
Molecular Formula: C6H3BCl2F3K
InChi Key: MASOPVJEJIORDH-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=C(Cl)C=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results