accessibility menu, dialog, popup

UserName

H3COOCH

CAS RN 1932-92-9

IUPAC Name: prop-2-yn-1-yl propanoate
Synonyms: prop-2-yn-1-yl propanoate
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: BGRYSGVIVVUJHH-UHFFFAOYSA-N
SMILES: CCC(=O)OCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results