accessibility menu, dialog, popup

UserName

H3COOSNOOCH3H3CCH3H2N

CAS RN 193537-14-3

IUPAC Name: 6-tert-butyl 3-ethyl 2-amino-4H,5H,6H,7H-thieno[2,3-c]pyridine-3,6-dicarboxylate
Synonyms: 6-tert-butyl 3-ethyl 2-amino-4H,5H,7H-thieno[2,3-c]pyridine-3,6-dicarboxylate
Molecular Weight (g/mol): 326.41
Molecular Formula: C15H22N2O4S
InChi Key: XSXVOVXVHBSSSN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC2=C1CCN(C2)C(=O)OC(C)(C)C
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results