accessibility menu, dialog, popup

UserName

NdClClCl

CAS RN 19423-78-0

IUPAC Name: trichloroneodymium hexahydrate
Synonyms: neodymium(III) chloride hexahydrate
Molecular Weight (g/mol): 358.68
Molecular Formula: Cl3H12NdO6
InChi Key: PCORICZHGNSNIZ-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.Cl[Nd](Cl)Cl
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)