accessibility menu, dialog, popup

UserName

H2NO

CAS RN 1947-47-3

IUPAC Name: 3-amino-2-phenyl-1H-inden-1-one
Synonyms: 3-amino-2-phenylinden-1-one
Molecular Weight (g/mol): 221.26
Molecular Formula: C15H11NO
InChi Key: HLEKBTIHDXNOQP-UHFFFAOYSA-N
SMILES: NC1=C(C(=O)C2=CC=CC=C12)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)