accessibility menu, dialog, popup

UserName

CAS RN 19493-44-8

ClN

CAS RN 19493-44-8

IUPAC Name: 1-chloroisoquinoline
Synonyms: 1-chloroisoquinoline
Molecular Weight (g/mol): 163.6
Molecular Formula: C9H6ClN
InChi Key: MSQCQINLJMEVNJ-UHFFFAOYSA-N
SMILES: ClC1=C2C=CC=CC2=CC=N1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)