CAS RN 19550-07-3
CAS RN 19550-07-3
IUPAC Name:
2,5-dimethylhexan-3-ol
Synonyms:
3-hexanol, 2,5-dimethyl-
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
SNKTZHPOKPYBPT-UHFFFAOYNA-N
SMILES:
CC(C)CC(O)C(C)C
1
–
1
of
1
results
1
2,5-Dimethyl-3-hexanol, 98%, Thermo Scientific™
CAS: 19550-07-3 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.231 MDL Number: MFCD00026452 InChI Key: SNKTZHPOKPYBPT-UHFFFAOYSA-N Synonym: 2,5-dimethyl-3-hexanol,3-hexanol, 2,5-dimethyl,acmc-209ezm,3-hexanol,2,5-dimethyl PubChem CID: 89183 IUPAC Name: 2,5-dimethylhexan-3-ol SMILES: CC(C)CC(C(C)C)O