accessibility menu, dialog, popup

UserName

CAS RN 19596-07-7

IUPAC Name: pent-4-ynenitrile
Synonyms: pent-4-ynenitrile
Molecular Weight (g/mol): 79.1
Molecular Formula: C5H5N
InChi Key: VMUWIDHKAIGONP-UHFFFAOYSA-N
SMILES: C#CCCC#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)