accessibility menu, dialog, popup

UserName

CAS RN 19598-90-4

H2OH2OH2OH2OH2OH2OGdONOOONOOONOO

CAS RN 19598-90-4

IUPAC Name: gadolinium(3+) hexahydrate trinitrate
Synonyms: gadolinium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 451.35
Molecular Formula: GdH12N3O15
InChi Key: XWFVFZQEDMDSET-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)