accessibility menu, dialog, popup

UserName

ONOOOOSOO

CAS RN 197244-91-0

IUPAC Name: (1,1-dioxo-1λ⁶-benzothiophen-2-yl)methyl 2,5-dioxopyrrolidin-1-yl carbonate
Synonyms: (1,1-dioxo-1λ⁶-benzothiophen-2-yl)methyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Weight (g/mol): 337.3
Molecular Formula: C14H11NO7S
InChi Key: XBVMGLSMCWWRLS-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC2=CC=CC=C2S1(=O)=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results