accessibility menu, dialog, popup

UserName

CAS RN 197304-21-5

H3COOOH3COOOH

CAS RN 197304-21-5

IUPAC Name: 4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid
Synonyms: 4-(4-formyl-3,5-dimethoxyphenoxy)butanoic acid
Molecular Weight (g/mol): 268.27
Molecular Formula: C13H16O6
InChi Key: SEJSVFHBVLKHLB-UHFFFAOYSA-N
SMILES: COC1=CC(OCCCC(O)=O)=CC(OC)=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results