CAS RN 197431-02-0
CAS RN 197431-02-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Desbutyl Dronedarone Hydrochloride, TRC
CAS: 197431-02-0 Molecular Formula: C27 H36 N2 O5 S . Cl H Molecular Weight (g/mol): 537.11 Synonym: Methanesulfonamide, N-[2-butyl-3-[4-[3-(butylamino)propoxy]benzoyl]-5-benzofuranyl]-, hydrochloride (1:1),Methanesulfonamide, N-[2-butyl-3-[4-[3-(butylamino)propoxy]benzoyl]-5-benzofuranyl]-, monohydrochloride (9CI),Desbutyldronedarone Hydrochloride,N-(2-Butyl-3-[4-[3-(butylamino)propoxy]benzoyl]-1-benzofuran-5-yl)methanesulfonamide hydrochloride IUPAC Name: N-[2-butyl-3-[4-[3-(butylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride SMILES: Cl.CCCCNCCCOc1ccc(cc1)C(=O)c2c(CCCC)oc3ccc(NS(=O)(=O)C)cc23