accessibility menu, dialog, popup

UserName

NaH3CCH3OO

CAS RN 19766-89-3

IUPAC Name: sodium 2-ethylhexanoate
Synonyms: sodium 2-ethylhexanoate
Molecular Weight (g/mol): 166.2
Molecular Formula: C8H15NaO2
InChi Key: VYPDUQYOLCLEGS-UHFFFAOYNA-M
SMILES: [Na+].CCCCC(CC)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)