CAS RN 19780-44-0
IUPAC Name:
4-ethylhexan-3-ol
Synonyms:
4-ethylhexan-3-ol
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
BOJLCKCCKQMGKD-UHFFFAOYNA-N
SMILES:
CCC(O)C(CC)CC
1
–
1
of
1
results
1
4-Ethyl-3-hexanol, 95%, Thermo Scientific™
CAS: 19780-44-0 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.231 MDL Number: MFCD00021917 InChI Key: BOJLCKCCKQMGKD-UHFFFAOYSA-N Synonym: 4-ethyl-3-hexanol,3-hexanol, 4-ethyl,3-ethyl hexanol-3,3-aethylhexanol-4,acmc-20amj6,4-ethyl-3-hexanol #,ethyl 3-pentyl carbinol PubChem CID: 140585 IUPAC Name: 4-ethylhexan-3-ol SMILES: CCC(CC)C(CC)O