accessibility menu, dialog, popup

UserName

CAS RN 197846-06-3

IUPAC Name: methyl 2-bromofuran-3-carboxylate
Synonyms: methyl 2-bromofuran-3-carboxylate
Molecular Weight (g/mol): 205.01
Molecular Formula: C6H5BrO3
InChi Key: XXUUZVIMSVBWIQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Br)OC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results