accessibility menu, dialog, popup

UserName

CH3OOCH3ON

CAS RN 19788-35-3

IUPAC Name: methyl 5-methyl-1,2-oxazole-3-carboxylate
Synonyms: methyl 5-methyl-1,2-oxazole-3-carboxylate
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: MVHHQOCEOUNTID-UHFFFAOYSA-N
SMILES: COC(=O)C1=NOC(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results