CAS RN 198022-70-7
CAS RN 198022-70-7
IUPAC Name:
N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
Synonyms:
N-(4-hydroxyphenyl)icosa-5,8,11,14-tetraenamide
Molecular Weight (g/mol):
395.59
Molecular Formula:
C26H37NO2
InChi Key:
IJBZOOZRAXHERC-UHFFFAOYSA-N
SMILES:
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NC1=CC=C(O)C=C1