accessibility menu, dialog, popup

UserName

CAS RN 1983205-53-3

IUPAC Name: potassium [4-(diethylcarbamoyl)phenyl]trifluoroboranuide
Synonyms: potassium [4-(diethylcarbamoyl)phenyl]trifluoroboranuide
Molecular Weight (g/mol): 283.14
Molecular Formula: C11H14BF3KNO
InChi Key: RILAOSSCNJEGBF-UHFFFAOYSA-N
SMILES: [K+].CCN(CC)C(=O)C1=CC=C(C=C1)[B-](F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results