accessibility menu, dialog, popup

UserName

NHNS

CAS RN 19933-24-5

IUPAC Name: 5-(thiophen-2-yl)-1H-pyrazole
Synonyms: 3-(thiophen-2-yl)-2H-pyrazole
Molecular Weight (g/mol): 150.2
Molecular Formula: C7H6N2S
InChi Key: TVNDPZYOQCCHTJ-UHFFFAOYSA-N
SMILES: N1N=CC=C1C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results