accessibility menu, dialog, popup

UserName

CH3NNNH2NH2OHSOOHO

CAS RN 20055-01-0

IUPAC Name: 1-methyl-1H-pyrazole-4,5-diamine; sulfuric acid
Synonyms: 2-methylpyrazole-3,4-diamine; sulfuric acid
Molecular Weight (g/mol): 210.21
Molecular Formula: C4H10N4O4S
InChi Key: CIMLJTZGZLWBJY-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.CN1N=CC(N)=C1N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results