accessibility menu, dialog, popup

UserName

CH3NHH3COO

CAS RN 2016-04-8

IUPAC Name: [bis(benzyloxy)methyl]dimethylazanium
Synonyms: [bis(benzyloxy)methyl]dimethylazanium
Molecular Weight (g/mol): 272.37
Molecular Formula: C17H22NO2
InChi Key: JFIKHFNGAURIIB-UHFFFAOYSA-O
SMILES: C[NH+](C)C(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results