accessibility menu, dialog, popup

UserName

CAS RN 2016-57-1

H3CNH2

CAS RN 2016-57-1

IUPAC Name: decan-1-amine
Synonyms: N-decylamine
Molecular Weight (g/mol): 157.3
Molecular Formula: C10H23N
InChi Key: MHZGKXUYDGKKIU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCN
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)