accessibility menu, dialog, popup

UserName

NaOOONN

CAS RN 20187-55-7

IUPAC Name: sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate
Synonyms: bendazac
Molecular Weight (g/mol): 304.28
Molecular Formula: C16H13N2NaO3
InChi Key: GHFGHIKJGVMEFT-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)