accessibility menu, dialog, popup

UserName

ONNH2

CAS RN 204078-48-8

IUPAC Name: 1-[2-(morpholin-4-yl)phenyl]methanamine
Synonyms: 1-[2-(morpholin-4-yl)phenyl]methanamine
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: RNYURNUANACIKS-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results