CAS RN 20412-39-9
CAS RN 20412-39-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methylbutyl Chloroformate, TRC
CAS: 20412-39-9 Molecular Formula: C6H11ClO2 Molecular Weight (g/mol): 150.6 Synonym: 2-Methyl-1-butanol Chloroformate,2-Methylbutyl Ester Carbonochloridic Acid IUPAC Name: 2-methylbutyl carbonochloridate SMILES: O=C(Cl)OCC(C)CC