CAS RN 2044-28-2
CAS RN 2044-28-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
2
2-Pyridylethylmercaptan, TRC
CAS: 2044-28-2 Molecular Formula: C7 H9 N S Molecular Weight (g/mol): 139.22 IUPAC Name: 2-pyridin-2-ylethanethiol SMILES: SCCc1ccccn1