accessibility menu, dialog, popup

UserName

H3COOHNClCl

CAS RN 2044-72-6

IUPAC Name: N-(2,5-dichlorophenyl)-3-oxobutanamide
Synonyms: N-(2,5-dichlorophenyl)-3-oxobutanamide
Molecular Weight (g/mol): 246.09
Molecular Formula: C10H9Cl2NO2
InChi Key: HLMZZYYGOKOOTA-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)NC1=CC(Cl)=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results