CAS RN 204634-20-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Tryptophan-15N2, TRC
CAS: 204634-20-8 Molecular Formula: C11H1215N2O2 Molecular Weight (g/mol): 206.21 Synonym: L-Tryptophan-15N2,L-[15N2]-Tryptophan,(S)-Tryptophan-15N2,(S)-α-(Amino-15N)-3-(indole-15N)propionic acid,(S)-α-Aminoindole-3-propanoic acid 15N2 IUPAC Name: (2S)-2-(^{15}N)azanyl-3-(1H-indol-3-yl)propanoic acid SMILES: [15NH2][C@H](C(O)=O)CC1=C[15N]([H])C2=C1C=CC=C2