accessibility menu, dialog, popup

UserName

H3CH3COOOHOOO

CAS RN 20513-98-8

IUPAC Name: 9-hydroxy-4,4-dimethyl-3,5,7,11-tetraoxatricyclo[6.3.0.0²,⁶]undecan-10-one
Synonyms: 9-hydroxy-4,4-dimethyl-3,5,7,11-tetraoxatricyclo[6.3.0.0²,⁶]undecan-10-one
Molecular Weight (g/mol): 216.19
Molecular Formula: C9H12O6
InChi Key: BDBGJSXZKMTMGP-UHFFFAOYNA-N
SMILES: CC1(C)OC2OC3C(O)C(=O)OC3C2O1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)