accessibility menu, dialog, popup

UserName

CAS RN 205692-63-3

OONNOO

CAS RN 205692-63-3

IUPAC Name: pyrazine-2,5-dicarboxylate
Synonyms: pyrazine-2,5-dicarboxylate
Molecular Weight (g/mol): 166.09
Molecular Formula: C6H2N2O4
InChi Key: GMIOYJQLNFNGPR-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=CN=C(C=N1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results