accessibility menu, dialog, popup

UserName

SOOH

CAS RN 2060-64-2

IUPAC Name: 1-benzothiophene-5-carboxylic acid
Synonyms: 1-benzothiophene-5-carboxylic acid
Molecular Weight (g/mol): 178.21
Molecular Formula: C9H6O2S
InChi Key: SNBYTKLWZRHESA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2SC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results